Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pth2r
Entrez Gene ID
81753
Gene Name
parathyroid hormone 2 receptor
Gene Aliases
Pthr2
Chromosome Location
Chr.9: 72052966 - 72157771 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031089.2 NP_112351.1 3 - 4 511 114
GenBank mRNA U55836.1 - 3 - 4 511 114

View Full Product Details