Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Thbd
Entrez Gene ID
83580
Gene Name
thrombomodulin
Gene Aliases
-
Chromosome Location
Chr.3: 142748673 - 142752325 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031771.2 NP_113959.1 1 - 1 1849 62
GenBank mRNA AF022743.1 - 1 - 1 1774 62
BC070903.1 - 1 - 1 1849 62

View Full Product Details