Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gldn
Entrez Gene ID
315675
Gene Name
gliomedin
Gene Aliases
Colm, Crgl2
Chromosome Location
Chr.8: 58870516 - 58914605 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181382.2 NP_852047.1 9 - 10 1349 104
GenBank mRNA AY266116.1 - 9 - 10 1349 104

View Full Product Details