Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc10a6
Entrez Gene ID
289459
Gene Name
solute carrier family 10 (sodium/bile acid cotransporter), member 6
Gene Aliases
Soat
Chromosome Location
Chr.14: 7618022 - 7682377 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198049.1 NP_932166.1 2 - 3 500 74
GenBank mRNA AJ583503.1 - 2 - 3 500 74

View Full Product Details