Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hnf1b
Entrez Gene ID
25640
Gene Name
HNF1 homeobox B
Gene Aliases
LF-B3, Tcf2, VHNF1
Chromosome Location
Chr.10: 71159984 - 71218607 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013103.1 NP_037235.1 6 - 7 1234 87
GenBank mRNA BC081826.1 - 5 - 6 1349 87
X56546.1 - 6 - 7 1234 87

View Full Product Details