Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myh7
Entrez Gene ID
29557
Gene Name
myosin, heavy chain 7, cardiac muscle, beta
Gene Aliases
Myhcb
Chromosome Location
Chr.15: 33634420 - 33657761 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017240.2 NP_058936.1 19 - 20 2432 115
XM_006251951.2 XP_006252013.1 21 - 22 2582 115
GenBank mRNA X15939.1 - 19 - 20 2434 115

View Full Product Details