Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psgb1
Entrez Gene ID
59313
Gene Name
pregnancy-specific beta 1-glycoprotein
Gene Aliases
Psg
Chromosome Location
Chr.1: 79181855 - 79190738 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021677.1 NP_067709.1 5 - 6 1277 143
XM_006228464.1 XP_006228526.1 4 - 5 1146 143
GenBank mRNA U66292.1 - 5 - 6 1277 143

View Full Product Details