Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdk2
Entrez Gene ID
81530
Gene Name
pyruvate dehydrogenase kinase, isozyme 2
Gene Aliases
-
Chromosome Location
Chr.10: 82838270 - 82852758 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030872.1 NP_110499.1 6 - 7 779 103
GenBank mRNA AF237719.1 - 6 - 7 779 103
BC061823.1 - 6 - 7 743 103
U10357.1 - 6 - 7 779 103

View Full Product Details