Assay Design
The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Gene Transcripts
- Gene Symbol
- Rpl36al
- Entrez Gene ID
-
81769
- Gene Name
- ribosomal protein L36a-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 91475222 - 91476540 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
M19635.1 |
- |
- |
41 |
71 |
View Full Product Details