The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031558.3 | NP_113746.1 | 3 - 4 | 422 | 79 |
GenBank mRNA | AB001349.1 | - | - | 378 | 79 |
AB006007.1 | - | 3 - 4 | 423 | 79 | |
AF044081.1 | - | - | 362 | 79 | |
AY736357.1 | - | 3 - 4 | 303 | 79 | |
BC088859.1 | - | 3 - 4 | 314 | 79 | |
EF091859.1 | - | 2 - 3 | 169 | 79 |