Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crybb3
Entrez Gene ID
64349
Gene Name
crystallin, beta B3
Gene Aliases
-
Chromosome Location
Chr.12: 49564996 - 49571006 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031690.1 NP_113878.1 1 - 2 114 111
XM_006249572.1 XP_006249634.1 2 - 3 152 111
GenBank mRNA AF287304.1 - 1 - 2 86 111
X05899.1 - 1 - 2 114 111

View Full Product Details