Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Deaf1
Entrez Gene ID
83632
Gene Name
DEAF1 transcription factor
Gene Aliases
-
Chromosome Location
Chr.1: 214283787 - 214317466 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031801.1 NP_113989.1 7 - 8 1100 132
XM_006230582.2 XP_006230644.1 7 - 8 1968 132
XM_006230583.2 XP_006230645.1 7 - 8 1968 132
XM_008760011.1 XP_008758233.1 7 - 8 1019 132
XM_008760012.1 XP_008758234.1 7 - 8 1019 132
XM_008760013.1 XP_008758235.1 7 - 8 1019 132
XM_008760014.1 XP_008758236.1 7 - 8 1984 132
XM_008760015.1 XP_008758237.1 2 - 3 612 132
XM_008760016.1 XP_008758238.1 2 - 3 612 132
GenBank mRNA AF055884.1 - 7 - 8 1100 132

View Full Product Details