Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdk4
Entrez Gene ID
89813
Gene Name
pyruvate dehydrogenase kinase, isozyme 4
Gene Aliases
-
Chromosome Location
Chr.4: 30546772 - 30556774 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053551.1 NP_446003.1 3 - 4 453 92
GenBank mRNA AF034577.1 - 3 - 4 453 92
FQ216954.1 - - 496 92

View Full Product Details