The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053584.2 | NP_446036.1 | 8 - 9 | 629 | 89 |
XM_006246914.1 | XP_006246976.1 | 8 - 9 | 697 | 89 | |
XM_008768132.1 | XP_008766354.1 | 7 - 8 | 500 | 89 | |
GenBank mRNA | BC126068.1 | - | 8 - 9 | 633 | 89 |