Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc17a7
Entrez Gene ID
116638
Gene Name
solute carrier family 17 (vesicular glutamate transporter), member 7
Gene Aliases
BNPI, Vglut1
Chromosome Location
Chr.1: 101161265 - 101172292 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053859.2 NP_446311.1 11 - 12 1514 94
GenBank mRNA EU253553.1 - 12 - 13 1479 94
FQ211811.1 - - 1527 94
U07609.1 - 11 - 12 1517 94

View Full Product Details