Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sec22a
Entrez Gene ID
117513
Gene Name
SEC22 vesicle trafficking protein homolog A (S. cerevisiae)
Gene Aliases
Sec22l2
Chromosome Location
Chr.11: 68632709 - 68686553 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057147.2 NP_476488.2 5 - 6 697 87
XM_006248419.2 XP_006248481.1 6 - 7 899 87
XM_006248420.2 XP_006248482.1 5 - 6 934 87
XM_008768692.1 XP_008766914.1 5 - 6 1146 87
XM_008768693.1 XP_008766915.1 5 - 6 1148 87
GenBank mRNA BC081746.1 - 5 - 6 697 87

View Full Product Details