Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cygb
Entrez Gene ID
170520
Gene Name
cytoglobin
Gene Aliases
Staap, Stap
Chromosome Location
Chr.10: 105618325 - 105628091 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130744.2 NP_570100.1 2 - 3 552 79
GenBank mRNA AJ245663.2 - 2 - 3 460 79
BC088455.1 - 2 - 3 552 79

View Full Product Details