Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spdya
Entrez Gene ID
192209
Gene Name
speedy/RINGO cell cycle regulator family member A
Gene Aliases
Gs4, Lm23, Spdy1, Spy1
Chromosome Location
Chr.6: 23498513 - 23543239 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138855.1 NP_620210.1 1 - 2 68 62
XM_006239768.2 XP_006239830.1 1 - 2 133 62
GenBank mRNA AF492385.1 - 1 - 2 68 62

View Full Product Details