Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Sycn
Entrez Gene ID
245917
Gene Name
syncollin
Gene Aliases
Syl
Chromosome Location
Chr.1: 85219827 - 85220239 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139086.1 NP_620786.1 1 - 1 46 72
GenBank mRNA AF008197.1 - 1 - 1 46 72

View Full Product Details