Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc15a4
Entrez Gene ID
246280
Gene Name
solute carrier family 15 (oligopeptide transporter), member 4
Gene Aliases
-
Chromosome Location
Chr.12: 32753824 - 32773277 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144758.1 NP_653359.1 5 - 6 1275 120
GenBank mRNA AB000280.1 - 5 - 6 1275 120
BC087709.1 - 5 - 6 1274 120

View Full Product Details