Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ms4a14
Entrez Gene ID
293743
Gene Name
membrane-spanning 4-domains, subfamily A, member 14
Gene Aliases
Sp3111
Chromosome Location
Chr.1: 227471441 - 227486953 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181082.2 NP_851596.1 1 - 2 256 123
GenBank mRNA AY258290.1 - 1 - 2 256 123

View Full Product Details