Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srr
Entrez Gene ID
303306
Gene Name
serine racemase
Gene Aliases
-
Chromosome Location
Chr.10: 61755213 - 61772332 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768015.1 XP_008766237.1 1 - 2 132 74
GenBank mRNA AB106282.1 - 1 - 2 84 74

View Full Product Details