Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam3c
Entrez Gene ID
312159
Gene Name
family with sequence similarity 3, member C
Gene Aliases
-
Chromosome Location
Chr.4: 49388962 - 49439892 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198771.1 NP_942066.1 7 - 8 513 106
GenBank mRNA AY228475.1 - 7 - 8 513 106

View Full Product Details