The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031553.2 | NP_113741.1 | 6 - 7 | 721 | 147 |
XM_006241196.2 | XP_006241258.1 | 6 - 7 | 739 | 147 | |
GenBank mRNA | BC089791.1 | - | 6 - 7 | 721 | 147 |
M55045.1 | - | 5 - 6 | 590 | 147 |