Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdv3
Entrez Gene ID
315970
Gene Name
carnitine deficiency-associated gene expressed in ventricle 3
Gene Aliases
-
Chromosome Location
Chr.8: 111837235 - 111850393 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014097.1 NP_001014119.1 4 - 5 1094 70
XM_008766529.1 XP_008764751.1 4 - 5 929 70
GenBank mRNA BC083654.1 - 4 - 5 1094 70

View Full Product Details