Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dalrd3
Entrez Gene ID
363146
Gene Name
DALR anticodon binding domain containing 3
Gene Aliases
-
Chromosome Location
Chr.8: 117355943 - 117358819 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014214.1 NP_001014236.1 5 - 6 930 60
GenBank mRNA BC082046.1 - 5 - 6 930 60

View Full Product Details