Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ckap2l
Entrez Gene ID
680515
Gene Name
cytoskeleton associated protein 2-like
Gene Aliases
-
Chromosome Location
Chr.3: 121796221 - 121822346 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006234995.2 XP_006235057.1 7 - 8 1892 82

View Full Product Details