Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Evx2
Entrez Gene ID
288155
Gene Name
even-skipped homeobox 2
Gene Aliases
-
Chromosome Location
Chr.3: 61564917 - 61581598 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_221512.8 XP_221512.5 1 - 2 428 107

View Full Product Details