Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mfap1a
Entrez Gene ID
499878
Gene Name
microfibrillar-associated protein 1A
Gene Aliases
RGD1562232
Chromosome Location
Chr.3: 113427229 - 113445458 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191964.1 NP_001178893.1 5 - 6 1010 71

View Full Product Details