Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vps18
Entrez Gene ID
296083
Gene Name
vacuolar protein sorting 18 homolog (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.3: 111097352 - 111108206 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106499.2 NP_001099969.1 2 - 3 490 97
GenBank mRNA BC169083.1 - 2 - 3 490 97

View Full Product Details