Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc24a5
Entrez Gene ID
311387
Gene Name
solute carrier family 24 (sodium/potassium/calcium exchanger), member 5
Gene Aliases
-
Chromosome Location
Chr.3: 117335212 - 117354480 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107769.1 NP_001101239.1 1 - 2 189 61

View Full Product Details