Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Apip
Entrez Gene ID
295961
Gene Name
APAF1 interacting protein
Gene Aliases
RGD1564562
Chromosome Location
Chr.3: 92969129 - 92998104 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106492.1 NP_001099962.1 6 - 7 743 127
XM_006234638.2 XP_006234700.1 6 - 7 792 127

View Full Product Details