The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173437.1 | NP_001166908.1 | 2 - 3 | 448 | 98 |
GenBank mRNA | EF688602.1 | - | 2 - 3 | 448 | 98 |