Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tgm6
Entrez Gene ID
296152
Gene Name
transglutaminase 6
Gene Aliases
RGD1565348
Chromosome Location
Chr.3: 122644183 - 122680049 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008762236.1 XP_008760458.1 1 - 2 1581 92

View Full Product Details