Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pld4
Entrez Gene ID
362792
Gene Name
phospholipase D family, member 4
Gene Aliases
-
Chromosome Location
Chr.6: 137323713 - 137331231 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126288.1 NP_001119760.1 1 - 2 129 73

View Full Product Details