Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Asb15
Entrez Gene ID
500050
Gene Name
ankyrin repeat and SOCS box containing 15
Gene Aliases
-
Chromosome Location
Chr.4: 51598829 - 51642276 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006236158.2 XP_006236220.1 3 - 4 511 90
XM_008762831.1 XP_008761053.1 3 - 4 667 90
XM_008762832.1 XP_008761054.1 3 - 4 468 90
XM_008762833.1 XP_008761055.1 3 - 4 689 90
GenBank mRNA BC097405.1 - 3 - 4 223 90

View Full Product Details