Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc35a3
Entrez Gene ID
310808
Gene Name
solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member A3
Gene Aliases
-
Chromosome Location
Chr.2: 219702681 - 219741886 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008761461.1 XP_008759683.1 1 - 2 388 65

View Full Product Details