The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006233199.2 | XP_006233261.1 | 7 - 8 | 1145 | 90 |
XM_006233200.2 | XP_006233262.1 | 7 - 8 | 1145 | 90 | |
XM_006233204.2 | XP_006233266.1 | 7 - 8 | 2508 | 90 | |
XM_008761446.1 | XP_008759668.1 | 8 - 9 | 1268 | 90 | |
GenBank mRNA | BC098000.1 | - | 7 - 8 | 1024 | 90 |