Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc39
Entrez Gene ID
691307
Gene Name
leucine rich repeat containing 39
Gene Aliases
-
Chromosome Location
Chr.2: 219598162 - 219617498 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109637.1 NP_001103107.1 2 - 3 345 109
XM_006233231.2 XP_006233293.1 2 - 3 332 109
GenBank mRNA BC158606.1 - 2 - 3 301 109
FQ215425.1 - 2 - 3 286 109

View Full Product Details