Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bmpr1b
Entrez Gene ID
310914
Gene Name
bone morphogenetic protein receptor, type IB
Gene Aliases
CFK-43a
Chromosome Location
Chr.2: 247403821 - 247676541 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006233400.2 XP_006233462.1 3 - 4 676 87
XM_006233401.2 XP_006233463.1 2 - 3 337 87
XM_008761513.1 XP_008759735.1 2 - 3 536 87
XM_008761514.1 XP_008759736.1 5 - 6 1058 87

View Full Product Details