Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrif1
Entrez Gene ID
310775
Gene Name
ligand dependent nuclear receptor interacting factor 1
Gene Aliases
RGD1306520
Chromosome Location
Chr.2: 209358656 - 209448123 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127485.1 NP_001120957.1 1 - 2 221 123

View Full Product Details