Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrif1
Entrez Gene ID
310775
Gene Name
ligand dependent nuclear receptor interacting factor 1
Gene Aliases
RGD1306520
Chromosome Location
Chr.2: 209358656 - 209448123 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006233149.2 XP_006233211.1 1 - 2 1515 135

View Full Product Details