Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ephx3
Entrez Gene ID
366836
Gene Name
epoxide hydrolase 3
Gene Aliases
Abhd9
Chromosome Location
Chr.7: 14212470 - 14217778 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108988.1 NP_001102458.1 2 - 3 519 73

View Full Product Details