Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Btbd2
Entrez Gene ID
500793
Gene Name
BTB (POZ) domain containing 2
Gene Aliases
RGD1566094
Chromosome Location
Chr.7: 11929891 - 11943559 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006241037.2 XP_006241099.2 6 - 7 1156 55
XM_008765152.1 XP_008763374.1 6 - 7 1174 55

View Full Product Details