Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mob3a
Entrez Gene ID
362833
Gene Name
MOB kinase activator 3A
Gene Aliases
Mobkl2a, Mobkl2b
Chromosome Location
Chr.7: 11878943 - 11896226 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108734.2 NP_001102204.1 3 - 4 691 70
XM_006240963.2 XP_006241025.1 3 - 4 1855 70
GenBank mRNA BC168845.1 - 3 - 4 691 70

View Full Product Details