The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127635.2 | NP_001121107.1 | 4 - 5 | 372 | 87 |
GenBank mRNA | BC166539.1 | - | 4 - 5 | 372 | 87 |