Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gdap1
Entrez Gene ID
312890
Gene Name
ganglioside-induced differentiation-associated-protein 1
Gene Aliases
-
Chromosome Location
Chr.5: 1328963 - 1347946 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107897.1 NP_001101367.1 2 - 3 404 80

View Full Product Details