The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127495.1 | NP_001120967.1 | 4 - 5 | 987 | 154 |
GenBank mRNA | BC160842.1 | - | 4 - 5 | 987 | 154 |