Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Terf1
Entrez Gene ID
297758
Gene Name
telomeric repeat binding factor (NIMA-interacting) 1
Gene Aliases
-
Chromosome Location
Chr.5: 2853455 - 2882728 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012464.1 NP_001012482.1 7 - 8 911 140
XM_006237729.2 XP_006237791.1 6 - 7 884 140
GenBank mRNA BC089888.1 - 7 - 8 911 140

View Full Product Details