Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3h12a
Entrez Gene ID
313587
Gene Name
zinc finger CCCH type containing 12A
Gene Aliases
RGD1306776
Chromosome Location
Chr.5: 143111339 - 143120131 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077671.1 NP_001071139.1 1 - 2 57 121
GenBank mRNA BC127516.1 - 1 - 2 57 121

View Full Product Details